Potassium nitrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

204110-50G 204110-250G
SIAL204110-50GEA 182 GBP
SIAL204110-50G SIAL204110-250G
Potassium nitrate, Sigma-Aldrich®
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR