Potassium iodide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

204102-10G 204102-250G 204102-50G
SIAL204102-10GEA 136 GBP
SIAL204102-10G SIAL204102-250G SIAL204102-50G
Potassium iodide, Sigma-Aldrich®
Potassium iodide
Formula: Kl
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 686 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR