Lead(II) iodide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

203602-50G
SIAL203602-50GEA 263 GBP
SIAL203602-50G
Lead(II) iodide, Sigma-Aldrich®
Lead (II) iodide
Formula: I₂Pb
Boiling Pt: 954 °C (1013 hPa)
Melting Pt: 402 °C
Density: 6.16 g/cm³ (20 °C)
MDL Number: MFCD00011163
CAS Number: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 12,05430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR