Cadmium(II) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

202908-10G 202908-50G
SIAL202908-10GEA 112 GBP
SIAL202908-10G SIAL202908-50G
Cadmium(II) chloride, Sigma-Aldrich®
Cadmium (II) chloride
Formula: CdCl₂
Boiling Pt: 967 °C (1013 hPa)
Melting Pt: 565 °C
Density: 4.05 g/cm³ (20 °C)
MDL Number: MFCD00010916
CAS Number: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 12,01653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR