L(+)-Lactic acid 85% ≥85% in aqueous solution, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

199257-1KG 199257-250G
SIAL199257-1KGEA 266 GBP
SIAL199257-1KG SIAL199257-250G
L(+)-Lactic acid 85% ≥85% in aqueous solution, Sigma-Aldrich®
L(+)-Lactic acid
Formula: C₃H₆O₃
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR