2-Chloropyrimidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

193291-10G 193291-50G
SIAL193291-10GEA 78.2 GBP
SIAL193291-10G SIAL193291-50G
2-Chloropyrimidine, Sigma-Aldrich®
2-Chloropyrimidine
Formula: C₄H₃ClN₂
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 65…66 °C
MDL Number: MFCD00006060
CAS Number: 1722-12-9
EINECS: 217-020-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR