2,3-Dihydroxybenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

189839-25G 189839-5G
SIAL189839-25GEA 143 GBP
SIAL189839-25G SIAL189839-5G
2,3-Dihydroxybenzaldehyde, Sigma-Aldrich®
2,3-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00003324
CAS Number: 24677-78-9
EINECS: 246-398-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR