(±)-4-Phenyl-2-butanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

189685-5G
SIAL189685-5GEA 76.8 GBP
SIAL189685-5G
(±)-4-Phenyl-2-butanol, Sigma-Aldrich®
(±)-4-Phenyl-2-butanol
Formula: (C₆H₅)CH₂CH₂CH(OH)CH₃
MW: 150.22 g/mol
Boiling Pt: 132 °C (1013 hPa)
Density: 0.9934 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044349
CAS Number: 2344-70-9
EINECS: 219-055-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR