Silicone oil, 60000 cSt, fluid, Dow Corning Corporation 200®, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Polyphenyl-methylsiloxane

M-Clarity™ quality level = MQ100

181838-1L 181838-250ML
SIAL181838-1LEA 250 GBP
SIAL181838-1L SIAL181838-250ML
Silicone oil, 60000 cSt, fluid, Dow Corning Corporation 200®, Sigma-Aldrich®
Silicone oil
Density: 0.98 g/mL (25 °C)
MDL Number: MFCD00132673
CAS Number: 63148-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR