1-Naphthyl isocyanate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Naphthylisocyanate

M-Clarity™ quality level = MQ100

170518-5G
SIAL170518-5GEA 38.8 GBP
SIAL170518-5G
1-Naphthyl isocyanate, Sigma-Aldrich®
1-Naphthyl isocyanate
Formula: C₁₁H₇NO
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 36 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00003881
CAS Number: 86-84-0
EINECS: 201-703-7
UN: 2206
ADR: 6.1,III
Merck Index: 12,06497

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR