Diethyl pyrocarbonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

M-Clarity™ quality level = MQ200

159220-100G 159220-25G 159220-5G
SIAL159220-100GEA 343 GBP
SIAL159220-100G SIAL159220-25G SIAL159220-5G
Diethyl pyrocarbonate, Sigma-Aldrich®
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
Boiling Pt: 93 °C (1013 hPa)
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR