Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Dibenzo-18-crown-6

M-Clarity™ quality level = MQ100

158399-10G 158399-2.5G 158399-50G
SIAL158399-10GEA 111 GBP
SIAL158399-10G SIAL158399-2.5G SIAL158399-50G
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I), Sigma-Aldrich®
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)
Formula: C₂₀H₂₄O₆
MDL Number: MFCD00005098
CAS Number: 14187-32-7
EINECS: 238-041-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR