1,3-Dibromo-5,5-dimethylhydantoin, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

157902-25G 157902-500G
SIAL157902-25GEA 49.2 GBP
SIAL157902-25G SIAL157902-500G
1,3-Dibromo-5,5-dimethylhydantoin, Sigma-Aldrich®
1,3-Dibromo-5,5-dimethylhydantoin
Formula: C₅H₆Br₂N₂O₂
MDL Number: MFCD00003189
CAS Number: 77-48-5
EINECS: 201-030-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR