Dimethyl suberate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Dimethyl octanedioate, 1,8-Octanedioic acid dimethyl ester, Suberic acid dimethyl ester

M-Clarity™ quality level = MQ100

149012-25G
SIAL149012-25GEA 86.5 GBP
SIAL149012-25G
Dimethyl suberate, Sigma-Aldrich®
Dimethyl suberate
Formula: C₁₀H₁₈O₄
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: –3.1 °C
Density: 1.014 g/cm³ (18 °C)
MDL Number: MFCD00008471
CAS Number: 1732-09-8
EINECS: 217-059-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR