2,4,5-Trimethoxybenzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

138894-10G
SIAL138894-10GEA 37.2 GBP
SIAL138894-10G
2,4,5-Trimethoxybenzoic acid, Sigma-Aldrich®
2,4,5-Trimethoxybenzoic acid
Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 142 °C
MDL Number: MFCD00002435
CAS Number: 490-64-2
EINECS: 207-715-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR