trans-2-Hexenal, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (E)-2-Hexenal, trans-2-Hexenal

M-Clarity™ quality level = MQ100

132659-25G 132659-5G
SIAL132659-25GEA 85.8 GBP
SIAL132659-25G SIAL132659-5G
trans-2-Hexenal, Sigma-Aldrich®
trans-2-Hexenal
Formula: C₆H₁₀O
Boiling Pt: 147 °C (1013 hPa)
Density: 0.846 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007008
CAS Number: 6728-26-3
EINECS: 229-778-1
UN: 1989
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR