4-Methylphenethylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

132020-1G 132020-5G
SIAL132020-1GEA 32.6 GBP
SIAL132020-1G SIAL132020-5G
4-Methylphenethylamine, Sigma-Aldrich®
4-Methylphenethylamine
Formula: C₉H₁₃N
MDL Number: MFCD00008195
CAS Number: 3261-62-9
EINECS: 221-865-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR