Cyclohexene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


MQ200

125431-2.5L 125431-500ML
SIAL125431-2.5LEA 199 GBP
SIAL125431-2.5L SIAL125431-500ML
Cyclohexene, Sigma-Aldrich®
Cyclohexene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 83
Melting Pt: –104 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00001539
CAS Number: 110-83-8
EINECS: 203-807-8
UN: 2256
ADR: 3 II
Merck Index: 13,02756

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR