3-Methyl-2-pentene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

111775-1G
SIAL111775-1GEA 112 GBP
SIAL111775-1G
3-Methyl-2-pentene, Sigma-Aldrich®
3-Methyl-2-pentene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 70…71 °C (1013 hPa)
Density: 0,6934 g/cm³ (20 °C)
MDL Number: MFCD00009334
CAS Number: 922-61-2
EINECS: 213-077-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR