Butyl vinyl ether, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: n-Butyl vinyl ether

M-Clarity™ quality level = MQ200

110299-100ML 110299-1L 110299-2L 110299-500ML
SIAL110299-100MLEA 38 GBP
SIAL110299-100ML SIAL110299-1L SIAL110299-2L SIAL110299-500ML
Butyl vinyl ether, Sigma-Aldrich®
Butyl vinyl ether
Formula: C₆H₁₂O
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –91.9 °C
Density: 0.7804 g/cm³ (20 °C)
MDL Number: MFCD00009454
CAS Number: 111-34-2
EINECS: 203-860-7
UN: 3271
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR