Chloroacetone stabilised, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Chloropropanone

10905-1KG
SIAL10905-1KGEA 192 GBP
SIAL10905-1KG
Chloroacetone stabilised, Sigma-Aldrich®
Chloroacetone

Packaging: produced by Wacker Chemie AG

Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 120
Melting Pt: –44 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 36
Storage Temperature: Refrigerator
MDL Number: MFCD00000936
CAS Number: 78-95-5
EINECS: 201-161-1
UN: 1695
ADR: 6.1 I
Merck Index: 13,02131

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR