α-Bromo-2,6-dichlorotoluene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ300

100307-25G
SIAL100307-25GEA 39.2 GBP
SIAL100307-25G
α-Bromo-2,6-dichlorotoluene, Sigma-Aldrich®
α-Bromo-2,6-dichlorotoluene
Formula: C₇H₅BrCl₂
Boiling Pt: 133…135 °C (10 mmHg)
Melting Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00000577
CAS Number: 20443-98-5
EINECS: 243-827-4
UN: 3261
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR