N-Ethylmaleimide (NEM), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione

M-Clarity™ quality level = MQ200

04259-25G 04259-5G
SIAL04259-25GEA 468 GBP
SIAL04259-25G SIAL04259-5G
N-Ethylmaleimide (NEM), Sigma-Aldrich®
N-Ethylmaleimide
Formula: C₆H₇NO₂
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 46 °C
MDL Number: MFCD00005509
CAS Number: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1,II
Merck Index: 13,03855

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR