Flame photometers, 420
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
47542000 47542100
SHEW47542000EA 7920 GBP
SHEW47542000 SHEW47542100
Flame photometers, 420
Spectrophotometers and Photometers Flame Photometers
Flame photometers designed for both sodium and potassium to be calibrated, measured and displayed simultaneously. The analytical performance is improved by the use of a lithium internal standard signal, which compensates for fluctuation in flame conditions, drift and dilution errors.
  • Fully automatic ignition and flame optimisation
  • Date and time of analysis is printed at each calibration
  • Quality check and repeat samples are identified on the printout
In 'Continuous' mode, it displays the current value on each channel and through the analogue ports. AnMore Product Information