Bisacrylamide

Supplier: IBI SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

IB70102
SHETIB70102EA 0 GBP
SHETIB70102
Bisacrylamide
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
Boiling Pt: 125 °C (3.33 hPa)
Melting Pt: 185 °C
Density: 1.235 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR