Human Recombinant IFNg (from E. coli)
Supplier: Shenandoah Biotechnology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100-77-100UG 100-77-1MG 100-77-20UG 100-77-250UG 100-77-500UG
SHBT100-77-100UGEA 172 GBP
SHBT100-77-100UG SHBT100-77-1MG SHBT100-77-20UG SHBT100-77-250UG SHBT100-77-500UG
Human Recombinant IFNg (from E. coli)
Proteins and Peptides
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Human IFN-γ is not cross-reactive with mouse IFN-γ.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.