Human Recombinant Oncostatin M (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100-56AF-100UG 100-56AF-10UG 100-56AF-1MG 100-56AF-500UG
SHBT100-56AF-100UGEA 728 GBP
SHBT100-56AF-100UG SHBT100-56AF-10UG SHBT100-56AF-1MG SHBT100-56AF-500UG
Human Recombinant Oncostatin M (Animal free) (from E. coli)
Proteins and Peptides
Oncostatin M is a cytokine that is produced by macrophages, dendritic cells, and T lymphocytes during inflammatory events. The Type-I and Type-II Oncostatin M receptors are located on the cell surface of endothelial and tumor cells, contain the glycoprotein 130 (gp130) subunit, and activate the JAK/STAT signaling pathway. Oncostatin M functions to inhibit tumor cell proliferation, induce liver stem cell maturation, regulate cytokine production during hematopoiesis and inflammation, stimulate bone formation, and promote nervous system development.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.