Human recombinant FGF21 (from E. coli)
Supplier: Shenandoah Biotechnology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100-148AF100UG 100-148AF-1MG 100-148AF-20UG 100-148AF500UG 100-148AF-5UG
SHBT100-148AF100UGEA 476 GBP
SHBT100-148AF100UG SHBT100-148AF-1MG SHBT100-148AF-20UG SHBT100-148AF500UG SHBT100-148AF-5UG
Human recombinant FGF21 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 21 (FGF-21) is an endocrine hormone that regulates energy homeostasis and exerts cardioprotective functions during heart injury. FGF-21 is expressed in the liver, pancreas, heart, and adipose tissues. FGF-21 signaling is activated through the FGFR1c receptor and β-Klotho co-receptor. FGF-21 is an important regulator of glucose uptake and reduces cell apoptosis under stress conditions.
  • Animal-free
  • Produced from genetically modified E. coli
  • Monomeric protein containing 182 amino acids
Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: This product is for research use only and is not for use in humans.