Cell culture medium, HyClone™ ActiPro
Supplier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH31037.01 SH31039.04 SH31037.08 SH31039.03
SH31037.01EA 445 GBP
SH31037.01 SH31039.04 SH31037.08 SH31039.03
Cell culture medium, HyClone™ ActiPro
Cell Culture Media
HyClone™ ActiPro cell culture medium has been formulated to provide high yields of recombinant proteins in processes using Chinese hamster ovary (CHO) cell lines.
  • Demonstrated high yields of recombinant proteins
  • Does not contain hypoxanthine and thymidine
  • Chemically defined ADCF formulation
  • Production medium for batch or fed-batch processes
  • Accompanied with detailed protocols for use to maximise output
HyClone™ ActiPro does not contain any growth factors such as insulin, peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batch consistency and increased cell culture process efficiency.