HyClone™ Bovine Growth Serum, US origin
Supplier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH30541.03 SH30541.02 SH30541.03HI SH30541.02HI SH30541.03IR25
HYCLSH30541.03EA 322 GBP
HYCLSH30541.03 HYCLSH30541.02 HYCLSH30541.03HI HYCLSH30541.02HI SH30541.03IR25
HyClone™ Bovine Growth Serum, US origin
Sera (Serum)
HyClone™ Bovine Growth Serum is a foetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Growth promotion studies demonstrate that BGS is an excellent replacement for FBS
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
HyClone Bovine Growth Serum, U.S. Origin is supplemented with chemically defined components including amino acids, traceMore Product Information