Fetal bovine serum characterized (United States), HyClone™
Supplier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH30071.03IH25 SH30071.03IH30
SH30071.03IH25EA 1080 GBP
SH30071.03IH25 SH30071.03IH30
Fetal bovine serum characterized (United States), HyClone™
Sera (Serum)
Enhance growth of cells with U.S.-sourced Cytiva HyClone™ characterised fetal bovine serum (FBS). More economical than defined FBS, but meets the requirements of most discriminating cell culturists.
Tested for endotoxin, hemoglobin, and additional biochemical assays and filtered through serial 100 nm pore-size rated filters.