Pure-Pass™ Well Plates, 96- and 384-Wells
Supplier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
451001 451002 451006 451007 451008 451009 455001 455002 455006 455007 455008 455009
SGEA451001EA 54.1 GBP
SGEA451001 SGEA451002 SGEA451006 SGEA451007 SGEA451008 SGEA451009 SGEA455001 SGEA455002 SGEA455006 SGEA455007 SGEA455008 SGEA455009
Pure-Pass™ Well Plates, 96- and 384-Wells
Microplates Deep Well Microplates
These as-molded, untreated plates are manufactured from a high purity polypropylene copolymer, making them some of the cleanest uncoated plastic plates.
Pure-Pass uncoated well plates are manufactured from a high clarity, high purity polypropylene copolymer for ultra-low extractables. These are molded in a cleanroon and meets all ANSI and SLAS standards.
Pure-Pass coated well plates with the addition of a glass barrier gives an added protection against extractables. Surface chemistry like glass could significantly reduce adsorption risk.