Advanced Green 3 Low-Bleed GC Septas
Supplier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
246510 246526 246527 246528 041916 041917 246124 246225 246228 246325 246424 246525 246624 246713 246725
SGEA246510EA 73.8 GBP
SGEA246510 SGEA246526 SGEA246527 SGEA246528 SGEA041916 SGEA041917 SGEA246124 SGEA246225 SGEA246228 SGEA246325 SGEA246424 SGEA246525 SGEA246624 SGEA246713 SGEA246725
Advanced Green 3 Low-Bleed GC Septas
Septa
Advanced Green 3 low-bleed GC septa is a premium septa for autosamplers in all critical analyses.
  • Made from low-bleed silicone
  • Excellent durability, resealing, solvent resistant, and tear resistant
  • Reliabily seal against the carrier gas pressure in the inlet
  • Capable of being pierced and resealing time after time without pieces of the septa being
  • deposited in the GC inlet system
  • Does not contaminate or bleed material into the chromatographic system
Combines significantly longer injection life, low bleed and low injection port adhesion.