SGE OptChem™ ConnecTite Inlet Liners
Supplier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0926232401 09262324 0926232501 09262325 0926232601 09262327
SGEA0926232401EA 0 GBP
SGEA0926232401 SGEA09262324 SGEA0926232501 SGEA09262325 SGEA0926232601 SGEA09262327
SGE OptChem™ ConnecTite Inlet Liners
Chromatography Supplies Chromatography Inlet Liners
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to go
  • Confidence in your analysis - certified deactivation
  • Robust and durable for longer life, less maintenance downtime and reduced maintenance costs
  • Color coded orange
  • Analytical formatted
ConnecTite liners facilitate maximum transfer of sample to the GC column and inhibit sample degradation by reducing contact with hot metal componentsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable