Tubing cutter, Terry-Tool
Supplier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
082780 082782
SGEA082780EA 272.8 GBP
SGEA082780 SGEA082782
Tubing cutter, Terry-Tool
Cutters
Cutter for steel tubing.
  • Cutting wheel for cutting ¹/₁₆ and ¹/₈" stainless steel tubing
  • Clean, right angle cuts with minimal burring or chipping of the tubing
  • No additional reaming or deburring is required
Ordering information: Spare cutting wheels are available.