Dialysis tubing, SERVAPOR®
Supplier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
44139.01 44139.02 44144.01 44144.02 44145.01 44145.04 44146.01 44146.04 44148.01 44148.02
SERA44139.01EA 45.92 GBP
SERA44139.01 SERA44139.02 SERA44144.01 SERA44144.02 SERA44145.01 SERA44145.04 SERA44146.01 SERA44146.04 SERA44148.01 SERA44148.02
Dialysis tubing, SERVAPOR®
Tubing Dialysis Tubing
Dialysis tubing is made of cellulose membrane. It contains low amounts of impurities such as sulphides and heavy metals.
  • High chemical and physical resistance
  • pH range: 2 to 12
  • Temperature range: 4 to 60 °C