Spermidine trihydrochloride

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Azoniaoctamethylenediammonium trichloride, 4-Azaoctamethylenediamine trihydrochloride, N1-(3-Aminopropyl)butane-1,4-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride

39786.01 39786.02
SERA39786.01EA 0 GBP
SERA39786.01 SERA39786.02
Spermidine trihydrochloride
Spermidine trihydrochloride
Formula: C₇H₂₂Cl₃N₃
MW: 254.63 g/mol
Melting Pt: 256…258 °C
Storage Temperature: Freezer
MDL Number: MFCD00012918
CAS Number: 334-50-9
EINECS: 206-379-0
Merck Index: 12,08893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR