2,3,5-Triphenyltetrazolium chloride (TTC)

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

37130.01 37130.02 37130.03
SERA37130.01EA 34.87 GBP
SERA37130.01 SERA37130.02 SERA37130.03
2,3,5-Triphenyltetrazolium chloride (TTC)
2,3,5-Triphenyltetrazolium chloride
Formula: C₁₉H₁₅ClN₄
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: 235 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011963
CAS Number: 298-96-4
EINECS: 206-071-6
Merck Index: 13,09815

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR