Silver nitrate

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

35110.01 35110.02 35110.03
SERA35110.01EA 61.23 GBP
SERA35110.01 SERA35110.02 SERA35110.03
Silver nitrate
Silver nitrate
Formula: AgNO₃
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR