4-Aminoantipyrine

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

12373.01
SERA12373.01EA 0 GBP
SERA12373.01
4-Aminoantipyrine
4-Aminoantipyrine
Formula: C₁₁H₁₃N₃O
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR