Buckets with Lid, Round
Supplier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2886 2887 4619 2884 4620
SEMA2886EA 611 GBP
SEMA2886 SEMA2887 SEMA4619 SEMA2884 SEMA4620
Buckets with Lid, Round
Buckets
Packing buckets, made of white PP, for industrial and laboratory applications. With handle and tight closing lid.
  • Stackable and sturdy with plastic carrying handle
  • Good chemical resistance
  • Easy to clean