Safety baskets
Supplier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2698 4954 2699 4953
216-7333EA 57.7 GBP
216-7333 SEMA4954 SEMA2699 SEMA4953
Safety baskets
Baskets Safety Transporting Baskets
PP, coloured.
  • Anti-static
  • Space-saving, stackable
  • Good chemical and temperature resistance
The antistatic safety baskets with perforated walls and base comes with collapsible plastic mounting bracket with lock in vertical position.