Storage Bags
Supplier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2452 2446 2455 2440 2442 2448 2439 2447 2450 4447
SEMA2452EA 51.5 GBP
SEMA2452 SEMA2446 SEMA2455 SEMA2440 SEMA2442 SEMA2448 SEMA2439 SEMA2447 SEMA2450 SEMA4447
Storage Bags
Bags Storage Bags
Storage bags in transparent LDPE, with bottom weld.
They are also suitable for liquids but without side weld.
Packaging: Minimum quantity:100 pieces