Bottles, wide neck, with screw cap
Supplier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0128
SEMA0128EA 82.2 GBP
SEMA0128
Bottles, wide neck, with screw cap
Bottles Plastic Bottles
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance