Bottle top acid dispenser, BASELINE®
Supplier: SEASTAR CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S077200-SSN002
SEACS077200-SSN002EA 0 GBP
SEACS077200-SSN002
Bottle top acid dispenser, BASELINE®
Bottle Top Dispensers
SEASTAR™ BASELINE® bottle-top dispenser, constructed from modified PTFE, combines the convenience of a traditional pump dispenser with the chemical compatibility and high purity.
  • Can be used with high purity concentrated acid
  • Wetted parts are extensively acid pre-cleaned
All wetted parts of the BASELINE® model have been extensively acid pre-cleaned parts and assembled in a trace metal cleanroom. Once assembled, the dispenser is then calibrated and mechanically tested to ensure the unit meets the required volume, precision and accuracy requirements.More Product Information