Exhaust filters, V3.0
Supplier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
407982 410535 490336 490914 410534 490335 407986 490986
SCTV407982EA 102 GBP
SCTV407982 SCTV410535 SCTV490336 SCTV490914 SCTV410534 SCTV490335 SCTV407986 SCTV490986
Exhaust filters, V3.0
Filters Venting Filters
The SCAT V3.0 exhaust filter for safety waste caps guarantees over 90% CTC adsorption for maximum air purity due to three integrated activated carbon types.
  • Blocks off harmful vapours and ensuring safe pressure equalisation in the waste containers
  • 3-layer filter media with high performance spectrum for acids and alkalis
  • Optimised for binding HPLC buffer solutions and absorbing organic solvent vapours
  • The natural pore structure of the activated carbon prevents sticking or clogging
  • Precise indication of the next replacement time by means of a time indicator
More Product Information