Safety funnels with lid, 'MARCO' V2.0, electrically conductive
Supplier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
317637 317651 318960 318961 318962 318964 318980 318981 318983 318984 318985 318988 306980 318630
221-0296EA 276 GBP
221-0296 SCTV317651 221-0299 221-0300 221-0301 SCTV318964 SCTV318980 SCTV318981 221-0302 SCTV318984 SCTV318985 SCTV318988 SCTV306980 SCTV318630
Safety funnels with lid, 'MARCO' V2.0, electrically conductive
Closures Safety Caps and Safety Waste Caps
S.C.A.T. 'MARCO' V2.0 safety funnels can be closed safely and prevent danger of ignition.
  • 'MARCO', V2.0 safety lid funnel is a modern lid funnel made of high-quality HDPE with extremely high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents explosive or health-endangering gases escaping
  • Sturdy hinge guarantees the closing mechanism has a long service life
More Product Information