Spillage trays
Supplier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
117983 117984 108981 117985 117986
590-0809EA 74.4 GBP
590-0809 SCTV117984 216-0924 216-0925 216-0926
Spillage trays
Trays/Dishes/Pots Spillage Trays/Pans
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
  • Electrically conductive available
  • Available with base insert; the canisters always stay dry
  • Anti-static mat and grund cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.