Self-adhesive labels, DURAN™ YOUTILITY
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
294010203
SCOT294010203EA 23.3 GBP
SCOT294010203
Self-adhesive labels, DURAN™ YOUTILITY
Labels
These self-adhesive, white polyester labels are designed for use with the DURAN™ YOUTILITY bottle system.
  • Chemical resistant
  • Temperature resistant at −40 to +150 °C
  • Non-toxic, and lead free
  • Autoclave/steam sterilisation Class 1 process indicator (ANSI/AAMI/ISO 11140-1:2005)
Surface will accept a variety of inks, including copier, printer, ballpoint and technical markers.