Pipette bulb, natural rubber, DURAN®

Supplier: DWK Life Sciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

292000205
SCOT292000205EA 14.4 GBP
SCOT292000205
Pipette bulb, natural rubber, DURAN®
Pipettes Pipette Bulbs
Designed for removing or dispensing small quantities of liquids from Pasteur pipettes or droppers. The combination of the Pasteur pipette and rubber bulb can be used as a teat pipette.

  • 5 mm opening
  • Maximum operating temperature is 80 °C

The flexible rubber teat is suitable for use with Pasteur pipettes or droppers as a pipetting aid to dispense liquids, remove liquids, or add drops of reagents.

Packaging: 10 pieces supplied per pack.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR